Tisk…

Dibenzofuran-2-ylboronic acid 99%

Dodavatel: Apollo Scientific

Synonyms: Dibenzo[b,d]furan-2-ylboronic acid

OR51007-25G
APOSOR51007-25GEA 2980 CZK
APOSOR51007-25G
Dibenzofuran-2-ylboronic acid 99%
Dibenzofuran-2-ylboronic acid
Vzorec: C₁₂H₉BO₃
MW: 212,009 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11183307
CAS číslo: 402936-15-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR