Tisk…

Methyl 4-(bromomethyl)phenylacetate 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(2-Methoxy-2-oxoethyl)benzyl bromide, Methyl 2-(4-(bromomethyl)phenyl)acetate, 4-(Bromomethyl)phenylacetic acid methyl ester

OR5099-5G OR5099-1G OR5099-10G
APOSOR5099-5GEA 4040 CZK
APOSOR5099-5G APOSOR5099-1G APOSOR5099-10G
Methyl 4-(bromomethyl)phenylacetate 98%
Methyl 4-(bromomethyl)phenylacetate
Vzorec: C₁₀H₁₁BrO₂
MW: 243,097 g/mol
Bod tání: 37…39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783030
CAS číslo: 7398-42-7
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR