5-Chloro-2-nitrophenylacetic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR50988-100MG
OR50988-1G
OR50988-250MG
APOSOR50988-100MGEA
622
CZK
APOSOR50988-100MG
APOSOR50988-1G
APOSOR50988-250MG
5-Chloro-2-nitrophenylacetic acid
5-Chloro-2-nitrophenylacetic acid
Vzorec:
C₈H₆ClNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD03206465 CAS číslo: 22908-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...