5-Bromo-2,4-dimethylbenzoic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
OR50986-1G
OR50986-250MG
APOSOR50986-1GEA
12010
CZK
APOSOR50986-1G
APOSOR50986-250MG
5-Bromo-2,4-dimethylbenzoic acid 99%
5-Bromo-2,4-dimethylbenzoic acid
Vzorec:
C₉H₉BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD24107212 CAS číslo: 842136-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...