3,6-Dibromo-9-methyl-9H-carbazole
Dodavatel: Apollo Scientific
|
Varování
|
OR50970-1G
OR50970-25G
OR50970-5G
APOSOR50970-1GEA
15540
CZK
APOSOR50970-1G
APOSOR50970-25G
APOSOR50970-5G
3,6-Dibromo-9-methyl-9H-carbazole
3,6-Dibromo-9-methyl-9H-carbazole
Vzorec:
C₁₃H₉Br₂N MW: 339.025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00218274 CAS číslo: 58246-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...