XPhos Pd G1
Dodavatel: Apollo Scientific
|
Varování
|
OR50938-100MG
OR50938-1G
OR50938-250MG
APOSOR50938-100MGEA
3310
CZK
APOSOR50938-100MG
APOSOR50938-1G
APOSOR50938-250MG
XPhos Pd G1
XPhos Pd G1
Vzorec:
C₃₃H₄₉P·C₈H₁₀ClNPd MW: 738,761 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13194128 CAS číslo: 1028206-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...