Methyl 3-bromo-2-cyanobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
OR50927-100MG
OR50927-1G
OR50927-250MG
APOSOR50927-100MGEA
787
CZK
APOSOR50927-100MG
APOSOR50927-1G
APOSOR50927-250MG
Methyl 3-bromo-2-cyanobenzoate
Methyl 3-bromo-2-cyanobenzoate
Vzorec:
C₉H₆BrNO₂ MW: 240,053 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398740 CAS číslo: 1261653-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...