Tisk…

Diethyl 2-bromo-5-chloroterephthalate 97%

Dodavatel: Apollo Scientific

OR50907-10G OR50907-1G OR50907-5G
APOSOR50907-10GEA 9330 CZK
APOSOR50907-10G APOSOR50907-1G APOSOR50907-5G
Diethyl 2-bromo-5-chloroterephthalate 97%
Diethyl 2-bromo-5-chloroterephthalate
Vzorec: C₁₂H₁₂BrClO₄
Storage Temperature: Refrigerator
MDL Number: MFCD12407164
CAS číslo: 340148-60-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR