Tisk…

N-(Phenylmethoxy)urea

Dodavatel: Apollo Scientific
Varování

OR50831-1G OR50831-250MG
APOSOR50831-1GEA 5430 CZK
APOSOR50831-1G APOSOR50831-250MG
N-(Phenylmethoxy)urea
N-(Phenylmethoxy)urea
Vzorec: C₈H₁₀N₂O₂
MW: 166,177 g/mol
Bod tání: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00047867
CAS číslo: 2048-50-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR