Tisk…

7-Chloro-2-methylimidazo[4,5-c]pyridine 95+%

Dodavatel: Apollo Scientific
Varování

OR500036-100MG OR500036-250MG
APOSOR500036-100MGEA 3230 CZK
APOSOR500036-100MG APOSOR500036-250MG
7-Chloro-2-methylimidazo[4,5-c]pyridine 95+%
7-Chloro-2-methylimidazo[4,5-c]pyridine
Vzorec: C₇H₆ClN₃
MW: 167,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834589
CAS číslo: 929074-44-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR