6-Amino-7-bromo-2,3-dihydro-1H-inden-1-one 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR500032-100MG
OR500032-250MG
APOSOR500032-100MGEA
2320
CZK
APOSOR500032-100MG
APOSOR500032-250MG
6-Amino-7-bromo-2,3-dihydro-1H-inden-1-one 95+%
6-Amino-7-bromo-2,3-dihydro-1H-inden-1-one
Vzorec:
C₉H₈BrNO MW: 226,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16249550 CAS číslo: 681246-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...