Tisk…

5-Bromo-2H-pyrazolo[3,4-b]pyridine 95+%

Dodavatel: Apollo Scientific
Varování

OR500031-10G OR500031-1G OR500031-5G
APOSOR500031-10GEA 0 CZK
APOSOR500031-10G APOSOR500031-1G APOSOR500031-5G
5-Bromo-2H-pyrazolo[3,4-b]pyridine 95+%
5-Bromo-2H-pyrazolo[3,4-b]pyridine
Vzorec: C₆H₄BrN₃
MW: 198,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27988163
CAS číslo: 1449693-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR