Tisk…

2-Chloro-8-methoxyquinoxaline 95+%

Dodavatel: Apollo Scientific
Varování

OR500025-100MG OR500025-250MG
APOSOR500025-100MGEA 5530 CZK
APOSOR500025-100MG APOSOR500025-250MG
2-Chloro-8-methoxyquinoxaline 95+%
2-Chloro-8-methoxyquinoxaline
Vzorec: C₉H₇ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD25368345
CAS číslo: 659729-70-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR