3-Bromo-4-chloro-5-methylpyridine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR500016-1G
OR500016-250MG
APOSOR500016-1GEA
12830
CZK
APOSOR500016-1G
APOSOR500016-250MG
3-Bromo-4-chloro-5-methylpyridine 95%
3-Bromo-4-chloro-5-methylpyridine
Vzorec:
C₆H₅BrClN MW: 206.468 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18415475 CAS číslo: 1261786-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...