Tisk…

4-Bromo-5-chloro-2-methylpyridine 95+%

Dodavatel: Apollo Scientific
Varování

OR500015-100MG OR500015-250MG
APOSOR500015-100MGEA 4400 CZK
APOSOR500015-100MG APOSOR500015-250MG
4-Bromo-5-chloro-2-methylpyridine 95+%
4-Bromo-5-chloro-2-methylpyridine
Vzorec: C₆H₅BrClN
Storage Temperature: Refrigerator
MDL Number: MFCD17014484
CAS číslo: 1211529-34-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR