Tisk…

1-(3-Chloro-2-methoxyphenyl)ethanone 95+%

Dodavatel: Apollo Scientific
Varování

OR500013-1G OR500013-250MG
APOSOR500013-1GEA 3640 CZK
APOSOR500013-1G APOSOR500013-250MG
1-(3-Chloro-2-methoxyphenyl)ethanone 95+%
1-(3-Chloro-2-methoxyphenyl)ethanone
Vzorec: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08753704
CAS číslo: 99585-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR