2-Bromo-6-formylbenzonitrile 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR500008-100MG
OR500008-250MG
APOSOR500008-100MGEA
1660
CZK
APOSOR500008-100MG
APOSOR500008-250MG
2-Bromo-6-formylbenzonitrile 95+%
2-Bromo-6-formylbenzonitrile
Vzorec:
C₈H₄BrNO MW: 210,027 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18390711 CAS číslo: 77532-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...