Tisk…

1,2-Diethynylbenzene

Dodavatel: Apollo Scientific

OR49074-250MG
APOSOR49074-250MGEA 2820 CZK
APOSOR49074-250MG
1,2-Diethynylbenzene
1,2-Diethynylbenzene
Vzorec: C₁₀H₆
MW: 126,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28164040
CAS číslo: 21792-52-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR