2-(4-Hydroxybenzoyl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR4859-5G
OR4859-25G
OR4859-100G
APOSOR4859-5GEA
622
CZK
APOSOR4859-5G
APOSOR4859-25G
APOSOR4859-100G
2-(4-Hydroxybenzoyl)benzoic acid
2-(4-Hydroxybenzoyl)benzoic acid
Vzorec:
C₁₄H₁₀O₄ MW: 242.227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016507 CAS číslo: 85-57-4 EINECS: 201-616-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...