2-Bromo-5-methylbenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR48273-1G
OR48273-250MG
OR48273-5G
APOSOR48273-1GEA
5200
CZK
APOSOR48273-1G
APOSOR48273-250MG
APOSOR48273-5G
2-Bromo-5-methylbenzenesulphonyl chloride
2-Bromo-5-methylbenzenesulphonyl chloride
Vzorec:
C₇H₆BrClO₂S MW: 269,543 g/mol Bod tání: 61…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD07783823 CAS číslo: 141113-98-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...