Tisk…

4-Bromo-2-chloro-5-nitrophenol 95%

Dodavatel: Apollo Scientific
Varování

OR48270-1G OR48270-250MG OR48270-5G
APOSOR48270-1GEA 16340 CZK
APOSOR48270-1G APOSOR48270-250MG APOSOR48270-5G
4-Bromo-2-chloro-5-nitrophenol 95%
4-Bromo-2-chloro-5-nitrophenol
Vzorec: C₆H₃BrClNO₃
MW: 252,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31700576
CAS číslo: 2091684-77-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR