Tisk…

Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate

Dodavatel: Apollo Scientific
Varování

OR48263-1G OR48263-25G OR48263-5G
APOSOR48263-1GEA 622 CZK
APOSOR48263-1G APOSOR48263-25G APOSOR48263-5G
Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate
Ethyl 4-[(3-chloro-4-methoxybenzyl)amino]-2-(methylthio)pyrimidine-5-carboxylate
Vzorec: C₁₆H₁₈ClN₃O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD17012695
CAS číslo: 330785-81-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR