Tisk…

2-Formylcinnamic acid 97%

Dodavatel: Apollo Scientific
Varování

OR4818-1G OR4818-5G
APOSOR4818-1GEA 3410 CZK
APOSOR4818-1G APOSOR4818-5G
2-Formylcinnamic acid 97%
2-Formylcinnamic acid
Vzorec: C₁₀H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD03840500
CAS číslo: 130036-17-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR