3-Chloroimidazo[1,2-a]pyridine-7-carbohydrazide
Dodavatel: Apollo Scientific
|
Varování
|
OR48092-1G
OR48092-250MG
APOSOR48092-1GEA
1330
CZK
APOSOR48092-1G
APOSOR48092-250MG
3-Chloroimidazo[1,2-a]pyridine-7-carbohydrazide
3-Chloroimidazo[1,2-a]pyridine-7-carbohydrazide
Vzorec:
C₈H₇ClN₄O MW: 210,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32111678 CAS číslo: 2391987-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...