2-(2-Methylsulphanylphenyl)pyrrolidine 95%
Dodavatel: Apollo Scientific
OR480888-1G
OR480888-5G
APOSOR480888-1GEA
6120
CZK
APOSOR480888-1G
APOSOR480888-5G
2-(2-Methylsulphanylphenyl)pyrrolidine 95%
2-(2-Methylsulphanylphenyl)pyrrolidine
Vzorec:
C₁₁H₁₅NS Storage Temperature: Ambient |
MDL Number:
MFCD02663494 CAS číslo: 383127-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...