Tisk…

Ethyl-4-bromo-3-methyl-but-2-enoate 90%

Dodavatel: Apollo Scientific

OR480880-1G OR480880-5G
APOSOR480880-1GEA 6120 CZK
APOSOR480880-1G APOSOR480880-5G
Ethyl-4-bromo-3-methyl-but-2-enoate 90%
Ethyl-4-bromo-3-methyl-but-2-enoate
Vzorec: C₇H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD07778473
CAS číslo: 26918-14-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR