Tisk…

2-Isocyanoethylbenzene 95%

Dodavatel: Apollo Scientific

OR480865-1G OR480865-5G
APOSOR480865-1GEA 23010 CZK
APOSOR480865-1G APOSOR480865-5G
2-Isocyanoethylbenzene 95%
2-Isocyanoethylbenzene
Vzorec: C₉H₉N
Storage Temperature: Ambient
MDL Number: MFCD02664670
CAS číslo: 59795-89-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR