Tisk…

2-Bromo-5-methylfuran 95%

Dodavatel: Apollo Scientific
Varování

OR480864-500MG OR480864-5G
APOSOR480864-500MGEA 0 CZK
APOSOR480864-500MG APOSOR480864-5G
2-Bromo-5-methylfuran 95%
2-Bromo-5-methylfuran
Vzorec: C₅H₅BrO
Storage Temperature: Ambient
MDL Number: MFCD11870159
CAS číslo: 123837-09-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR