Tisk…

3-Methylpyrrolidin-3-ol 95%

Dodavatel: Apollo Scientific
Varování

OR480853-1G OR480853-5G
APOSOR480853-1GEA 1660 CZK
APOSOR480853-1G APOSOR480853-5G
3-Methylpyrrolidin-3-ol 95%
3-Methylpyrrolidin-3-ol
Vzorec: C₅H₁₁NO
MW: 101,147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100984
CAS číslo: 125032-87-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR