Tisk…

3-(Bromomethyl)-2-methylpyridine hydrobromide 95%

Dodavatel: Apollo Scientific

OR480851-1G OR480851-500MG
APOSOR480851-1GEA 16040 CZK
APOSOR480851-1G APOSOR480851-500MG
3-(Bromomethyl)-2-methylpyridine hydrobromide 95%
3-(Bromomethyl)-2-methylpyridine hydrobromide
Vzorec: C₇H₈BrN·BrH
MW: 266.961 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22200150
CAS číslo: 76915-53-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR