Tisk…

Methyl 4-(4-methylbenzoyl)benzoate 95%

Dodavatel: Apollo Scientific

OR480846-1G OR480846-5G
APOSOR480846-1GEA 3440 CZK
APOSOR480846-1G APOSOR480846-5G
Methyl 4-(4-methylbenzoyl)benzoate 95%
Methyl 4-(4-methylbenzoyl)benzoate
Vzorec: C₁₆H₁₄O₃
MW: 254,281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072893
CAS číslo: 64141-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR