Tisk…

1H-Benzimidazole-2-carboxylic acid methyl ester 95%

Dodavatel: Apollo Scientific

Synonyms: Methyl 1H-benzo[d]imidazole-2-carboxylate

OR480814-5G
APOSOR480814-5GEA 3970 CZK
APOSOR480814-5G
1H-Benzimidazole-2-carboxylic acid methyl ester 95%
1H-Benzimidazole-2-carboxylic acid methyl ester
Vzorec: C₉H₈N₂O₂
MW: 176,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03413579
CAS číslo: 5805-53-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR