Tisk…

Ethyl 2-bromo-2-cyclopropylacetate 90%, tech

Dodavatel: Apollo Scientific

OR480806-1G OR480806-100MG OR480806-250MG OR480806-500MG OR480806-2.5G
APOSOR480806-1GEA 4470 CZK
APOSOR480806-1G APOSOR480806-100MG APOSOR480806-250MG APOSOR480806-500MG APOSOR480806-2.5G
Ethyl 2-bromo-2-cyclopropylacetate 90%, tech
Ethyl 2-bromo-2-cyclopropylacetate
Vzorec: C₇H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD18837719
CAS číslo: 1200828-74-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR