4-Bromo-3-methylthiophene-2-carboxaldehyde 97%
Dodavatel: Apollo Scientific
OR480793-1G
OR480793-5G
APOSOR480793-1GEA
5130
CZK
APOSOR480793-1G
APOSOR480793-5G
4-Bromo-3-methylthiophene-2-carboxaldehyde 97%
4-Bromo-3-methylthiophene-2-carboxaldehyde
Vzorec:
C₆H₅BrOS MW: 205,072 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01859870 CAS číslo: 30153-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...