6-Bromo-8-methylquinoline 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR480792-1G
OR480792-5G
APOSOR480792-1GEA
2030
CZK
APOSOR480792-1G
APOSOR480792-5G
6-Bromo-8-methylquinoline 97%
6-Bromo-8-methylquinoline
Vzorec:
C₁₀H₈BrN Storage Temperature: Ambient |
MDL Number:
MFCD11847622 CAS číslo: 178396-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...