5-(Methoxymethyl)furan-2-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR480781-5G
APOSOR480781-5GEA
3770
CZK
APOSOR480781-5G
5-(Methoxymethyl)furan-2-carbaldehyde 95%
5-(Methoxymethyl)furan-2-carbaldehyde
Vzorec:
C₇H₈O₃ MW: 140,137 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02253954 CAS číslo: 1917-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...