Tisk…

3-Chloro-4-iodophenacyl bromide 95%

Dodavatel: Apollo Scientific

OR48077-1G OR48077-250MG
APOSOR48077-1GEA 18620 CZK
APOSOR48077-1G APOSOR48077-250MG
3-Chloro-4-iodophenacyl bromide 95%
3-Chloro-4-iodophenacyl bromide
Vzorec: C₈H₅BrClIO
MW: 359,386 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491127
CAS číslo: 1701815-08-8
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR