Sodium 5-hydroxypentanoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR48075-1G
OR48075-5G
APOSOR48075-1GEA
612
CZK
APOSOR48075-1G
APOSOR48075-5G
Sodium 5-hydroxypentanoate 95%
Sodium 5-hydroxypentanoate
Vzorec:
C₅H₉O₃·Na Storage Temperature: Ambient |
MDL Number:
MFCD22192345 CAS číslo: 37435-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...