Tisk…

7-Bromo-5-methyl-1H-indole-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR480741-1G OR480741-5G
APOSOR480741-1GEA 8370 CZK
APOSOR480741-1G APOSOR480741-5G
7-Bromo-5-methyl-1H-indole-2-carboxylic acid 95%
7-Bromo-5-methyl-1H-indole-2-carboxylic acid
Vzorec: C₁₀H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD02664426
CAS číslo: 15936-75-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR