4-Bromo-3,5-dimethoxybenzyl alcohol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(4-Bromo-3,5-dimethoxyphenyl)methanol
OR4806-100G
OR4806-25G
APOSOR4806-100GEA
8440
CZK
APOSOR4806-100G
APOSOR4806-25G
4-Bromo-3,5-dimethoxybenzyl alcohol 98%
4-Bromo-3,5-dimethoxybenzyl alcohol
Vzorec:
C₉H₁₁BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD06658150 CAS číslo: 61367-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...