Tisk…

2-Amino-2-(3-methoxyphenyl)acetic acid

Dodavatel: Apollo Scientific

OR480473-10G OR480473-5G
APOSOR480473-10GEA 23280 CZK
APOSOR480473-10G APOSOR480473-5G
2-Amino-2-(3-methoxyphenyl)acetic acid
2-Amino-2-(3-methoxyphenyl)acetic acid
Vzorec: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD02662407
CAS číslo: 7314-43-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR