Tisk…

2-Amino-2-(4-iodophenyl)acetic acid

Dodavatel: Apollo Scientific

OR480461-1G OR480461-2G OR480461-5G OR480461-100MG OR480461-250MG
APOSOR480461-1GEA 29960 CZK
APOSOR480461-1G APOSOR480461-2G APOSOR480461-5G APOSOR480461-100MG APOSOR480461-250MG
2-Amino-2-(4-iodophenyl)acetic acid
2-Amino-2-(4-iodophenyl)acetic acid
Vzorec: C₈H₈INO₂
MW: 277.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02662474
CAS číslo: 299167-68-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR