2-Amino-2-(1,3-benzodioxol-5-yl)acetic acid
Dodavatel: Apollo Scientific
OR480460-1G
OR480460-2G
OR480460-5G
APOSOR480460-1GEA
4860
CZK
APOSOR480460-1G
APOSOR480460-2G
APOSOR480460-5G
2-Amino-2-(1,3-benzodioxol-5-yl)acetic acid
2-Amino-2-(1,3-benzodioxol-5-yl)acetic acid
Vzorec:
C₉H₉NO₄ MW: 195,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00858979 CAS číslo: 39533-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...