2-Amino-2-(3-chloro-4-methoxy-phenyl)acetic acid
Dodavatel: Apollo Scientific
OR480451-1G
OR480451-2G
OR480451-5G
APOSOR480451-1GEA
4860
CZK
APOSOR480451-1G
APOSOR480451-2G
APOSOR480451-5G
2-Amino-2-(3-chloro-4-methoxy-phenyl)acetic acid
2-Amino-2-(3-chloro-4-methoxy-phenyl)acetic acid
Vzorec:
C₉H₁₀ClNO₃ MW: 215.634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02662433 CAS číslo: 53091-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...