Tisk…

2-Amino-2-(2,4,5-trimethoxyphenyl)acetic acid

Dodavatel: Apollo Scientific

OR480444-1G OR480444-2G
APOSOR480444-1GEA 4860 CZK
APOSOR480444-1G APOSOR480444-2G
2-Amino-2-(2,4,5-trimethoxyphenyl)acetic acid
2-Amino-2-(2,4,5-trimethoxyphenyl)acetic acid
Vzorec: C₁₁H₁₅NO₅
Storage Temperature: Ambient
MDL Number: MFCD02662391
CAS číslo: 318270-07-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR