(E)-3-Butoxy-1-(4-(methylthio)phenyl)prop-2-en-1-one
Dodavatel: Apollo Scientific
OR480290-1G
OR480290-250MG
OR480290-500MG
APOSOR480290-1GEA
21890
CZK
APOSOR480290-1G
APOSOR480290-250MG
APOSOR480290-500MG
(E)-3-Butoxy-1-(4-(methylthio)phenyl)prop-2-en-1-one
(E)-3-Butoxy-1-(4-(methylthio)phenyl)prop-2-en-1-one
Vzorec:
C₁₄H₁₈O₂S MW: 250,357 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22683242 CAS číslo: 1375008-08-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...