4-Amino-3-hydroxypiperidine 95%
Dodavatel: Apollo Scientific
OR480288-1G
OR480288-500MG
APOSOR480288-1GEA
10620
CZK
APOSOR480288-1G
APOSOR480288-500MG
4-Amino-3-hydroxypiperidine 95%
4-Amino-3-hydroxypiperidine
Vzorec:
C₅H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD22533520 CAS číslo: 1391004-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...