6-Methoxynaphthalene-2-sulphonyl chloride
Dodavatel: Apollo Scientific
OR480278-100MG
OR480278-1G
OR480278-500MG
APOSOR480278-100MGEA
4340
CZK
APOSOR480278-100MG
APOSOR480278-1G
APOSOR480278-500MG
6-Methoxynaphthalene-2-sulphonyl chloride
6-Methoxynaphthalene-2-sulphonyl chloride
Vzorec:
C₁₁H₉ClO₃S Storage Temperature: Ambient |
MDL Number:
MFCD12402643 CAS číslo: 56875-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...