Tisk…

8-Methoxy-1,2,3,4-tetrahydroisoquinoline 95%

Dodavatel: Apollo Scientific

OR480263-100MG OR480263-1G OR480263-250MG
APOSOR480263-100MGEA 4670 CZK
APOSOR480263-100MG APOSOR480263-1G APOSOR480263-250MG
8-Methoxy-1,2,3,4-tetrahydroisoquinoline 95%
8-Methoxy-1,2,3,4-tetrahydroisoquinoline
Vzorec: C₁₀H₁₃NO
MW: 163,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042690
CAS číslo: 34146-68-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR