Tisk…

4-(2H-1,2,3-Triazol-2-yl)piperidine 95%

Dodavatel: Apollo Scientific

OR480262-1G OR480262-250MG OR480262-500MG
APOSOR480262-1GEA 28830 CZK
APOSOR480262-1G APOSOR480262-250MG APOSOR480262-500MG
4-(2H-1,2,3-Triazol-2-yl)piperidine 95%
4-(2H-1,2,3-Triazol-2-yl)piperidine
Vzorec: C₇H₁₂N₄
Storage Temperature: Ambient
MDL Number: MFCD11042726
CAS číslo: 765270-45-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR